Learn More
WDR77 Mouse anti-Human, Mouse, Clone: OTI5H9, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809827

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles.Specifications
WDR77 | |
Monoclonal | |
Unconjugated | |
WDR77 | |
HKMT1069, MEP-50, MEP50, Nbla10071, p44, p44/Mep50 | |
Mouse | |
Affinity Chromatography | |
RUO | |
70465, 79084 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5H9 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q99J09, Q9BQA1 | |
Wdr77 | |
Human recombit protein fragment corresponding to amino acids 1-250 of human WDR77 produced in E.coli. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.