Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR77 Recombinant Rabbit Monoclonal Antibody (PSH0-07)
GREENER_CHOICE

Catalog No. FEMA550769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550769 Supplier Invitrogen™ Supplier No. MA550769
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Human prostate tissue, human fallopian tube tissue, mouse brain tissue, HepG2 cell lysate, 293 cell lysate, K562 cell lysate, Hela, HepG2. Predicted band size: 37 kDa Subcellular Location: Nucleus, Cytoplasm.

WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR77
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone PSH0-07
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene WDR77
Gene Accession No. Q9BQA1
Gene Alias 2610003I18Rik; 2610312E17Rik; Ac2-269; androgen receptor cofactor p44; C79984; HKMT1069; MEP50; MEP-50; Methylosome protein 50; MGC2722; Nbla10071; OTTHUMP00000013473; p44; p44/Mep50; RGD1310479; RP11-552M11.3; testis tissue sperm-binding protein Li 44a; WD repeat domain 77; WD repeat-containing protein 77; WD45; WDR77
Gene Symbols WDR77
Host Species Rabbit
Immunogen Recombinant protein within human WDR77 aa 1-342/342.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79084
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.