Learn More
WDR79 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
| Antigen | WDR79 |
|---|---|
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2264
|
Bethyl Laboratories, Inc
IHC00534T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2263
|
Bethyl Laboratories, Inc
IHC00534 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Monkey, Gorilla, Chimpanzee, Crab-eating macaque and White-tufted-ear marmoset
This gene encodes an essential component of the telomerase holoenzyme complex, a ribonucleoprotein complex required for telomere synthesis. This protein is enriched in Cajal bodies, nuclear sites of RNP processing that are important for telomerase function. It interacts with dyskerin, TERT and TERC, other components of active telomerase, and with small Cajal body RNAs (scaRNAs), which are involved in modifying splicing RNAs.Specifications
| WDR79 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| TCAB1, WDR79 | |
| WRAP53 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
| Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q9BUR4 | |
| 55135 | |
| Between 1 and 50 | |
| Primary | |
| 4° C | |
| WRAP53 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.