Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555455
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555455 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555455 Supplier Invitrogen™ Supplier No. PA555455
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVPVSLQTLK PVTDRANPKI GIGMLAFSPD SYFLATRNDN IPNAVWVWDI QKLRLFAVLE QLSPVRAFQW DPQQPRLAIC TGGSRLYLWS PAGCMSVQVP GEGDFAVLSL CWHLSGDSM Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 53%.

This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Studies of the related mouse protein suggest that the encoded protein may play a role in the process of ossification.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR8
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WRAP73
Gene Accession No. Q9P2S5
Gene Alias 2610044M17Rik; 5330425N03Rik; Dd57; LOW QUALITY PROTEIN: WD repeat-containing protein WRAP73; WD repeat containing, antisense to TP73; WD repeat containing, antisense to Trp73; WD repeat domain 8; WD repeat-containing protein 8; WD repeat-containing protein 8; WD repeat-containing protein WRAP73; WD repeat-containing protein antisense to TP73; WD repeat-containing protein antisense to TP73 gene; WD repeat-containing protein WRAP73; WDR8; WRAP73
Gene Symbols WRAP73
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR8. Recombinant protein control fragment (Product #RP-88840).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 49856
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.