Learn More
Invitrogen™ WDR82 Polyclonal Antibody

Description
Predicted to react with xenopus and chicken based on sequence homology.
Specifications
Specifications
| Antigen | WDR82 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | WDR82 |
| Gene Accession No. | Q6NV31, Q6UXN9, Q8BFQ4 |
| Gene Alias | 9430077D24Rik; Cdw5; CDW5/WDR82; MST107; MSTP107; PRO2730; PRO34047; Protein TMEM113; Protein word of mouth; PTK9L; Swd2; TMEM113; transmembrane protein 113; UNQ9342/PRO34047; WD repeat domain 82; WD repeat domain containing 82; WD repeat-containing protein 82; Wdr82; wdr82 protein; WDR82A; wom; word of mouth; wu:fb22h06; zgc:55453; zgc:85635; zgc:91817 |
| Gene Symbols | WDR82 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.