Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR82 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583499
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583499 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583499 Supplier Invitrogen™ Supplier No. PA583499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GYANSKAVTL EASFTPDSQF IMIGSEDGKI HVWNGESGIK VAVLDGKHTG PITCLQFNPK FMTFASACSN MAFWLPTIDD Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.

WDR82 is a component of the mammalian SET1A (MIM 611052)/SET1B (MIM 611055) histone H3-Lys4 methyltransferase complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR82
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR82
Gene Accession No. Q6UXN9
Gene Alias 9430077D24Rik; Cdw5; CDW5/WDR82; MST107; MSTP107; PRO2730; PRO34047; Protein TMEM113; Protein word of mouth; PTK9L; Swd2; TMEM113; transmembrane protein 113; UNQ9342/PRO34047; WD repeat domain 82; WD repeat domain containing 82; WD repeat-containing protein 82; Wdr82; wdr82 protein; WDR82A; wom; word of mouth; wu:fb22h06; zgc:55453; zgc:85635; zgc:91817
Gene Symbols WDR82
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR82. Recombinant protein control fragment (Product #RP-104719).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80335
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.