Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR83OS Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566788
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566788 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566788 Supplier Invitrogen™ Supplier No. PA566788
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSTNNMSDPRR PNKVLRYKPP PSECNPA Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

WDR83OS is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR83OS
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR83OS
Gene Accession No. Q9Y284
Gene Alias C19orf56; CGI-140; My006; Protein Asterix; PTD008; UPF0139 membrane protein C19orf56; WD repeat domain 83 opposite strand; WDR83 opposite strand; WDR83OS
Gene Symbols WDR83OS
Host Species Rabbit
Immunogen Recombinant Human WDR83OS. Recombinant protein control fragment (Product #RP-105363).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51398
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.