Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR85 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554632
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554632 Supplier Invitrogen™ Supplier No. PA554632
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WSWLLFRSLQ RAPSWSFPSN LGTKTADLKG ASELPTPCHE CREDNDGEGH ARPQSGMKPL TEGMRKNGTW LQATAATTRD CGVNPEEADS A Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 34%.

WDR85 is a WD repeat-containing protein that plays a role in the first step of diphthamide biosynthesis (Carette et al., 2009 [PubMed 19965467]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR85
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPH7
Gene Accession No. Q9BTV6
Gene Alias C9orf112; diphthamide biosynthesis 7; diphthamide biosynthesis protein 7; Diphthine methyltransferase; DPH7; RP11-48C7.3; RRT2; WD repeat domain 85; WD repeat-containing protein 85; WDR85
Gene Symbols DPH7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR85. Recombinant protein control fragment (Product #RP-93016).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92715
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.