Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR89 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551457 Supplier Invitrogen™ Supplier No. PA551457
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DEGFYWWDLN HLDTDEPVTR LNIQDVREVV NMKEDALDYL IGGLYHEKTD TLHVIGGTNK GRIHLMNCSM SGLTHVTSLQ GGHAATVRSF CWNVQDDSLL TGGEDAQLLL WKPGAIEKTF TKKESMKIAS SVHQRVRVHS N Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 79%.

WDR89 bears six WD repeats and is a member of the WD repeat family. The WD repeat is defined by four or more repeating units of a conserved core of approximately 40 amino acids ending with tryptophan-aspartic acid (WD). WD repeats may serve as sites of protein-protein interaction for adaptor proteins and facilitate multiprotein complex formation. The cellular function of WDR89 has not been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR89
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR89
Gene Accession No. Q96FK6
Gene Alias C14orf150; MSTP050; WD repeat domain 89; WD repeat-containing protein 89; WDR89
Gene Symbols WDR89
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR89. Recombinant protein control fragment (Product #RP-88901).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112840
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.