Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23816925UL
Description
WDR9 Polyclonal specifically detects WDR9 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
WDR9 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q9NSI6 | |
BRWD1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: TKVRTCMHNQKDAVQMPSETLKAKMVPEKVPRRCATVAANKIKIMSNLKETISGPENVWIRKSSRKLPHRN | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
bromodomain and WD repeat domain containing 1, bromodomain and WD repeat-containing protein 1, C21orf107, chromosome 21 open reading frame 107, FLJ11315, FLJ43918, N143, transcriptional unit N143, WD repeat domain 9, WD repeat protein WDR9-form2, WD repeat-containing protein 9, WDR9 | |
Rabbit | |
Affinity Purified | |
RUO | |
54014 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction