Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR91 Polyclonal Antibody

WDR91 Polyclonal Antibody

Supplier:  Thermo Scientific PA550863

Catalog No. PA550863


Only null left
Add to Cart

Description

Description

WDR91 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

WDR91
Polyclonal
Unconjugated
WDR91
9530020G05Rik; AI987683; AU018665; HSPC049; RGD1306697; SORF1; SORF-1; WD repeat domain 91; WD repeat-containing protein 91; Wdr91
Rabbit
Antigen Affinity Chromatography
RUO
29062
-20°C
Liquid
Immunohistochemistry (Paraffin)
1.4 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
A4D1P6
WDR91
Synthetic peptide of human WD repeat domain 91.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.