Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR92 Monoclonal Antibody (OTI2F2)
GREENER_CHOICE

Catalog No. PIMA527174
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527174 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527174 Supplier Invitrogen™ Supplier No. MA527174
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

WDR92 Monoclonal Antibody for Western Blot

WD40 repeats are a common structural module in eukaryotic proteins, and proteins containing WD40 domains have a wide range of functions, including signal transduction, cell cycle regulation, RNA splicing, and transcription. One such protein, WDR92, also known as monad, contains two WD40 repeats is widely expressed in human tissues, especially testis. Overexpression of this protein or its binding partner RNA polymerase II-associated protein 3 (RPAP3) potentiated apoptosis and caspase-3 activation induced by TNF-a and cycloheximide, suggesting that WDR92, together with RPAP3, may function as a novel modulator of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR92
Applications Western Blot
Classification Monoclonal
Clone OTI2F2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene WDR92
Gene Accession No. Q96MX6
Gene Alias AI553587; HZGJ; monad; RGD1563863; testicular secretory protein Li 67; WD repeat domain 92; WD repeat-containing protein 92; WD repeat-containing protein Monad; Wdr92
Gene Symbols WDR92
Host Species Mouse
Immunogen Full length human recombinant protein of WDR92 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116143
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.