Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDR97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584416 Supplier Invitrogen™ Supplier No. PA584416
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LMSYFLYSPV HCRPELKKLL HGLGLQDPEG FLFKEMMTWV QGPDLDSKAG LRTCCHQKLE DMIQELQETP SQTSVVSGAP TRASVIPSGT Highest antigen sequence indentity to the following orthologs: Mouse - 48%, Rat - 44%.

The function and protein-coding potential of this gene is unknown. The exon combination is based on AB058778.1 for which two possible open reading frames can be predicted (with start codons at nucleotide 26 or 2614). The position of the first ORF stop codon is consistent with a prediction of nonsense-mediated decay. Given the observation of the first ORF and its length, a predicted translation of the second ORF is inconsistent with the translation leaky scanning theory. Therefore, this gene is represented as a non-protein-coding transcript. [provided by RefSeq, Oct 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR97
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDR97
Gene Accession No. A6NE52
Gene Alias KIAA1875; WD repeat domain 97; WD repeat-containing protein 97; WDR97
Gene Symbols WDR97
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDR97. Recombinant protein control fragment (Product #RP-101969).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 340390
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.