Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDSOF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560732
Description
Immunogen sequence: GIGTRFCGTS FFTVGDDKTV KQWKMDGPGY GDEEEPLHTI LGKTVYTGID HHWKEAVFAT CGQQVDIWDE QRTNPICSMT WGFD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.
Possible role in ribosomal RNA processing. May function as a substrate receptor for CUL4-DDB1 E3 ubiquitin-protein ligase complex.
Specifications
WDSOF1 | |
Polyclonal | |
Unconjugated | |
DCAF13 | |
Dcaf13; DDB1 and CUL4 associated factor 13; DDB1- and CUL4-associated factor 13; Gm83; HSPC064; WD repeat and SOF domain-containing protein 1; WD repeats and SOF domain containing 1; WD repeats and SOF1 domain containing; WDSOF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25879 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NV06 | |
DCAF13 | |
Recombinant protein corresponding to Human WDSOF1. Recombinant protein control fragment (Product #RP-93878). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction