Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDSUB1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155073
Description
WDSUB1 Polyclonal specifically detects WDSUB1 in Human samples. It is validated for Western Blot.Specifications
WDSUB1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ36175,2610014F08Rik, SAM and U-box domain-containing protein 1, UBOX6, WD repeat, SAM and U-box domain containing 1, WD repeat, sterile alpha motif and U-box domain containing 1, WDSAM1 | |
Rabbit | |
53 kDa | |
100 μL | |
Primary | |
Goat: 83%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Zebrafish | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q8N9V3 | |
WDSUB1 | |
Synthetic peptides corresponding to WDSUB1(WD repeat, sterile alpha motif and U-box domain containing 1) The peptide sequence was selected from the middle region of WDSUB1. Peptide sequence KVKSLSSGIPDEFICPITRELMKDPVIASDGYSYEKEAMENWISKKKRTS The peptide sequence for this immunogen was taken from within the described region. | |
Affinity purified | |
RUO | |
151525 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction