Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDSUB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566143
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566143 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566143 Supplier Invitrogen™ Supplier No. PA566143
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IEELRTKVKSL SSGIPDEFIC PITRELMKDP VIASDGYSYE KEAMENWISK KKRTSPMTNL VLPSAVLTPN RTLKMAINRW LETHQ Highest antigen sequence identity to the following orthologs - mouse 90%, rat 90%.

WD repeat-containing proteins are those that contain 4 or more copies of the WD-repeat (tryptophan-aspartate repeat), a sequence motif approximately 31 amino acids long, that encodes a structural repeat. There are two isoforms of WDSUB1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDSUB1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDSUB1
Gene Accession No. Q8N9V3
Gene Alias 1700048E19Rik; 2610014F08Rik; UBOX6; WD repeat and SAM domain containing 1; WD repeat, SAM and U-box domain containing 1; WD repeat, SAM and U-box domain-containing protein 1; WD repeat, sterile alpha motif and U-box domain containing 1; WDSAM1; Wdsub1
Gene Symbols WDSUB1
Host Species Rabbit
Immunogen Recombinant Human WDSUB1. Recombinant protein control fragment (Product #RP-106825).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.