Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDSUB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567587
Description
WDSUB1 Polyclonal Antibody for Western Blot
WD repeat-containing proteins are those that contain 4 or more copies of the WD-repeat (tryptophan-aspartate repeat), a sequence motif approximately 31 amino acids long, that encodes a structural repeat. There are two isoforms of WDSUB1.
Specifications
WDSUB1 | |
Polyclonal | |
Unconjugated | |
WDSUB1 | |
1700048E19Rik; 2610014F08Rik; UBOX6; WD repeat and SAM domain containing 1; WD repeat, SAM and U-box domain containing 1; WD repeat, SAM and U-box domain-containing protein 1; WD repeat, sterile alpha motif and U-box domain containing 1; WDSAM1; Wdsub1 | |
Rabbit | |
affinity chromatography | |
RUO | |
151525, 72137 | |
-20°C | |
Liquid |
Western Blot | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8N9V3, Q9D0I6 | |
WDSUB1 | |
Synthesized peptide derived from human WDSUB1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction