Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WDTC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555785
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555785 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555785 Supplier Invitrogen™ Supplier No. PA555785
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDQHTIVWDP LHHKKLLSMH TGHTANIFSV KFLPHAGDRI LITGAADSKV HVHDLTVKET IHMFGDHTNR VKRIATAPMW PNTFWSAAE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen WDTC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WDTC1
Gene Accession No. Q8N5D0
Gene Alias adipose; adipose homolog; ADP; DCAF9; DDB1 and CUL4 associated factor 9; Gm695; KIAA1037; WD and tetratricopeptide repeats 1; WD and tetratricopeptide repeats protein 1; Wdtc1
Gene Symbols WDTC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WDTC1. Recombinant protein control fragment (Product #RP-94261).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23038
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.