Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDTC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113155
Description
WDTC1 Polyclonal Antibody for Western Blot, ELISA

Specifications
WDTC1 | |
Polyclonal | |
Unconjugated | |
WDTC1 | |
adipose; adipose homolog; ADP; DCAF9; DDB1 and CUL4 associated factor 9; Gm695; KIAA1037; WD and tetratricopeptide repeats 1; WD and tetratricopeptide repeats protein 1; Wdtc1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23038, 230796 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
0.75 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q80ZK9, Q8N5D0 | |
WDTC1 | |
Recombinant Human WD and tetratricopeptide repeats protein 1 protein (201-420AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction