Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ WDYHV1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565814
Description
Immunogen sequence: RPGDGPVIWDY HVVLLHVSSG GQNFIYDLDT VLPFPCLFDT YVEDAFKSDD DIHPQFRRKF RVIRADS Highest antigen sequence identity to the following orthologs - mouse 82%, rat 84%.
WDYHV1 mediates the side-chain deamidation of N-terminal glutamine residues to glutamate, an important step in N-end rule pathway of protein degradation. Conversion of the resulting N-terminal glutamine to glutamate renders the protein susceptible to arginylation, polyubiquitination and degradation as specified by the N-end rule. Does not act on substrates with internal or C-terminal glutamine and does not act on non-glutamine residues in any position. Does not deaminate acetylated N-terminal glutamine. With the exception of proline, all tested second-position residues on substrate peptides do not greatly influence the activity. In contrast, a proline at position 2, virtually abolishes deamidation of N-terminal glutamine.
Specifications
WDYHV1 | |
Polyclonal | |
Unconjugated | |
WDYHV1 | |
2410187C16Rik; AU014961; AW550036; C8orf32; nt(Q)-amidase; Ntaq1; N-terminal Gln amidase; protein NH2-terminal glutamine deamidase; Protein N-terminal glutamine amidohydrolase; WDYHV motif containing 1; WDYHV motif-containing protein 1; WDYHV1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55093 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96HA8 | |
WDYHV1 | |
Recombinant Human WDYHV1. Recombinant protein control fragment (Product #RP-106487). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction