Learn More
Description
- Super resilient PVC surface with PVC sponge designed for comfort and long life
- With vinyl surface and safety edges beveled with 1/2 in. overall thickness
- Available in Black (BK) or Black w/Yellow Borders (BYL)
Specifications
Specifications
| For Use With (Application) | Dry areas |
| Thickness (English) | 0.5 in. |
| Color | Black/Yellow |
| Dimensions (L x W) | 2 x 3 ft. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.