Learn More
Wearwell™ Grade A No. 482 Food Production Mats
Catalog No. 191500076 Shop All Wearwell Tennessee Mat Company Inc. Products
Click to view available options
Dimensions (L x W):
30 x 36 in.
30 x 60 in.
First antifatigue mat designed specifically for food production plants
  • Specially formulated nitrile rubber composite is 100% anti-microbial and withstands animal fats, fish and vegetable oils, and compounds used in a wide variety of food processing
  • Built-in handles ease cleaning of mats
  • Hygenic porthole and solid underside design simplify cleaning and virtually eliminate areas where food and bacteria can collect
  • Substantial underside knobs for increased comfort
  • Textured surface increases traction when greasy and wet
  • Terra cotta
Compliance
Meet NSF antimicrobial test ASTM™ G21-96 (2002).
Specifications
Dimensions (L x W) 30 x 60 in.
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.