Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wearwell™ Industrial WorkSafe™ Mat

Catalog No. p-101881
Click to view available options
Dimensions (L x W):
40 x 28 in.
64 x 40 in.

Extra-thick workstation mats withstand many industrial chemicals

  • Designed for metal working and other rough industrial areas where performance and productivity count
  • These extra thick and extra buoyant mats provide long-term comfort
  • The CFR (mineral oil cutting fluid resistant) compound withstands many non-petroleum based industrial fluids
  • The bright yellow or orange, molded-in safety borders define workstations while the mats textured surface adds traction in wet areas
  • In hazardously slippery areas, use the GritWorks version This high performance carbide grit coating offers 60% more traction than specified by OSHA in wet areas
  • 0.875 in. thick

Specifications

Dimensions (L x W) 64 x 40 in.
Color Black/Orange
Resistance Grease-resistant
Thickness (English) 0.87 in.
Material Rubber
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.