Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wearwell™ Ortho Stand™ Mats

Catalog No. p-101850
Click to view available options
Dimensions (L x W):
3 x 2 ft.
3 x 4 ft.
Color:
Black
Black with Yellow Border

Domed anti fatigue mat increases circulation

  • An economical, dry area, single work area mat is ergonomically designed to increase worker comfort
  • Hollow domed surface encourages subtle changes in posture increasing circulation and lessening fatigue
  • ideal for checkout stands, manifest stations, mailing areas and punch presses

Order Info

Overall thickness: 0.5 in. (1.3cm)

Specifications

Dimensions (L x W) 3 x 2 ft.
Color Black with Yellow Border
Material Natural Rubber

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.