Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wearwell™ Standard Corrugated Runner
Shop All Wearwell Tennessee Mat Company Inc. Products
Click to view available options
Dimensions (L x W):
105 x 2 ft.
105 x 3 ft.
105 x 4 ft.
Description
- Use in any indoor area requiring nonslip, traction-grip runner to protect floors
- Easy to clean
- Made of chemical resistant, 0.125 in. (0.32cm) thick vinly
Order Info
Available in cut lengths up to 150 ft. in 2 ft., 3 ft. and 4 ft. widths. Contact your Fisher Scientific™ Customer Service representative for ordering information.
Specifications
Specifications
| Dimensions (L x W) | 105 x 2 ft. |
| Material | Chemical-resistant Vinyl |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction