Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wearwell™ Diamond-Plate Modular Mat Systems

Catalog No. 19034142 Shop All Wearwell Tennessee Mat Company Inc. Products
Click to view available options
Resistance:
Grease-resistant
Greaseproof

Interlocking system ideal for use in single work spaces or large areas

  • Designed for comfort and ease of use in mostly dry areas
  • Interlocking system requires no additional bevels or attachments
  • Lightweight, easy to clean
  • Rounded exterior edges
  • Natural rubber for use in general applications
  • Grease-proof, 50% nitrile rubber for use in areas with occasional exposure to chemicals or caustic agents

Order Info

Thickness: 0.5 in. (1.3cm), dimensions: L x W: 3 x 3 ft. (91.4 x 91.4cm).

Specifications

Dimensions (L x W) 3 x 3 ft.
Resistance Greaseproof
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.