Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
American Educational Products Weather Symbol Clings

Click to view available options
Grade:
K to 4
Description
Tell the weather story on windows, wet erase boards, or even refrigerators! These 19 colorful static cling weather symbols are meteorologically accurate and easy to handle for even the smallest hands.
Specifications
Specifications
Includes | 2 Suns, 1 partlysunny/cloudy, 1 cloud, 1 stormclouds, 2 rains, 1 snow, 2 highs, 2 lows 2 cold fronts, 2 warm fronts, 1 tornado, 1 hurricane, 1 tropicalstorm |
Product Type | Weather Symbol Clings |
For Use With (Application) | Classroom maps |
Grade | K to 4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction