Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
AquaPhoenix™ Weather Wonders Deluxe Kit Components
Shop All Fisherbrand Products

Click to view available options
Product Type:
Cotton Swabs
Craft Stick
Deli Container
Fine Sand
Latch top Storage Container
Zipseal bag
Description
Variety of components specially formatted as replacements for AquaPhoenix custom designed hands-on educational kits.
Specifications
Specifications
Product Type | Fine Sand |
Grade | K to 5 |
For Use With (Application) | Education Kit |
Quantity | 4 cup |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction