Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WEE1 Polyclonal Antibody
WEE1 Polyclonal Antibody
Supplier: Thermo Scientific PA567019
Description
WEE1 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
WEE1 | |
Polyclonal | |
Unconjugated | |
WEE1 | |
CTRCT30; DKFZp686I18166; epididymis luminal protein 113; fa20h03; FLJ16446; HEL113; VIM; Vimentin; wee 1 homolog; WEE 1 homolog 1 (S. pombe); Wee1; WEE1 G2 checkpoint kinase; WEE1 homolog; wee1 protein; WEE1 tyrosine kinase; WEE1+ homolog; Wee1A; wee1A kinase; WEE1hu; wee1-like protein kinase; wu:fa20h03; zgc:136514 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7465 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.10 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P30291 | |
WEE1 | |
Recombinant Human WEE1. Recombinant protein control fragment (Product #RP-105459). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction