Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WEE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567588 Supplier Invitrogen™ Supplier No. PA567588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse and Rat samples.

WEE1, which is classified as a Ser/Thr kinase by sequence analysis, has been shown to inactivate multiple CDKs by tyrosine phosphorylation. Wee1 protein catalyzes the inhibitory tyrosine phosphorylation of CDC2/cyclin B kinase, and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WEE1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene WEE1
Gene Accession No. P30291, P47810, Q63802
Gene Alias CTRCT30; DKFZp686I18166; epididymis luminal protein 113; fa20h03; FLJ16446; HEL113; VIM; Vimentin; wee 1 homolog; WEE 1 homolog 1 (S. pombe); Wee1; WEE1 G2 checkpoint kinase; WEE1 homolog; wee1 protein; WEE1 tyrosine kinase; WEE1+ homolog; Wee1A; wee1A kinase; WEE1hu; wee1-like protein kinase; wu:fa20h03; zgc:136514
Gene Symbols WEE1
Host Species Rabbit
Immunogen Synthesized peptide derived from human WEE1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22390, 308937, 7465
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.