Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ WEE2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5140078
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5140078 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5140078 Supplier Invitrogen™ Supplier No. PA5140078
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: DIDKELRQKL NFSYCEETEI EGQKKVEESR EASSQTPEKG EVQDSEAKGT PPWTPLSNVH ELDTSSEKDK ESPDQILRTP VSHPLKC Highest antigen sequence indentity to the following orthologs: Rat - 41%, Mouse - 43%.

Oocyte-specific protein tyrosine kinase that phosphorylates and inhibits CDK1/CDC2 and acts as a key regulator of meiosis during both prophase I and metaphase II (PubMed:29606300). Required to maintain meiotic arrest in oocytes during the germinal vesicle (GV) stage, a long period of quiescence at dictyate prophase I, by phosphorylating CDK1 at 'Tyr-15', leading to inhibit CDK1 activity and prevent meiotic reentry. Also required for metaphase II exit during egg activation by phosphorylating CDK1 at 'Tyr-15', to ensure exit from meiosis in oocytes and promote pronuclear formation. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen WEE2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene WEE2
Gene Accession No. P0C1S8
Gene Alias Gm1065; mWee1B; WEE1 homolog 2 (S. pombe); WEE1B; Wee1B kinase; wee1-like protein kinase 1B; Wee1-like protein kinase 2; Wee2
Gene Symbols WEE2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human WEE2. Recombinant protein control fragment (Product #RP-108910).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 494551
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.