Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WEE2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548699
Description
Oocyte-specific protein tyrosine kinase that phosphorylates and inhibits CDK1 and acts as a key regulator of meiosis during both prophase I and metaphase II. Required to maintain meiotic arrest in oocytes during the germinal vesicle (GV) stage, a long period of quiescence at dictyate prophase I, by phosphorylating CDK1 at ′Tyr-15′, leading to inhibit CDK1 activity and prevent meiotic reentry. Also required for metaphase II exit during egg activation by phosphorylating CDK1 at ′Tyr-15′, to ensure exit from meiosis in oocytes and promote pronuclear formation.
Specifications
Wee2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P0C1S8, Q66JT0 | |
WEE2 | |
KLH conjugated synthetic peptide between 364-393 amino acids from the Central region of mouse Wee2. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
WEE2 | |
Wee1-like protein kinase 2, Wee1-like protein kinase 1B, Wee1B kinase, mWee1B, Wee2, Wee1b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
381759, 494551 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction