Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Welch™ Vacuum Pump On/Off Foot Switch

Vacuum pump accessory that provides hands-free operation of Welch DryFast diaphragm and WOB-L piston pumps.

$321.00 - $483.00

Specifications

Type Foot Switch
View More Specs

Products 3
Catalog Number Mfr. No. Electrical Requirements Model Price Quantity & Availability  
Catalog Number Mfr. No. Electrical Requirements Model Price Quantity & Availability  
01-055-16
View Documents
Welch™
1430B
115 V 60 Hz Momentary On/Off switch logic
Each for $483.00
Only null left
Add to Cart
 
01-055-17
View Documents
Welch™
1430C
90 to 230 V Momentary On/Off switch logic
Each for $454.00
Only null left
Add to Cart
 
01-104-17
View Documents
Welch™
1430A
115 V 60 Hz Maintained On/Off switch logic
Each for $321.00
Only null left
Add to Cart
 
Description

Description

  • Hands-free on/off pump operation
  • No adaptors or special connection needed
  • Plugs directly into the power source outlet
  • Switch logic and power varies depending on model:
    • Parts 01-104-17 and 01-055-16: 3-prong U.S. standard plug with 8 ft. cord (CSA, NEMA, and UL Enclosure Type 1)
    • Part 01-055-17: Male and female IEC connections with 8 ft. cord (plugs directly into the IEC connection on the pump)
Specifications

Specifications

Foot Switch
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.