Learn More
Restek™ Weldments for EZ Twist Top Split/Splitless Injection Ports
SDP
Catalog No. 06611233 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5890/6850/6890/7890 GCs
Agilent 6850/6890/7820 GCs
Agilent 6850/6890/7820 GCs (Large Canister Type Filter)
Material:
Siltek-Treated
Stainless Steel
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Material
06-611-233 Agilent 5890/6850/6890/7890 GCs Stainless Steel
06-611-241 Agilent 6850/6890/7820 GCs (Large Canister Type Filter) Siltek-Treated
06-611-240 Agilent 6850/6890/7820 GCs (Large Canister Type Filter) Stainless Steel
06-611-232 Agilent 6850/6890/7820 GCs Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-611-233 Supplier Restek™ Supplier No. 22724
Add to Cart
Add to Cart
For Agilent GCs
Specifications
Type Weldment
Material Stainless Steel
Includes 2 weldment O-rings installed on the weldment
For Use With (Application) EZ Twist Top Split/Splitless Injection Ports
Quantity 1/Pk.
For Use With (Equipment) Agilent 5890/6850/6890/7890 GCs