Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Polyurethane Coated Seamless Knit HPPE Gloves

Catalog Number 19148714
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

FlexTech™ Polyurethane Coated Seamless Knit HPPE Gloves are constructed with High Performance Polyethylene (HPPE) Fibers for increased cut protection. Polyurethane coating on the palm provides a non-slip surface for enhanced grip capabilities.

  • High performance polyethylene fibers (HPPE) blended with 13-gauge nylon for increased cut resistance
  • Gloves combine comfort, dexterity, and cut resistance
  • Polyurethane coating on the palm enhances grip and tactility
  • Launderable for an extended useful life
ANSI Abrasion Rating ANSI 6
Coated Area Palm, Fingers
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Hand Specific
Material HPPE, Nylon
Product Type Seamless Knit Gloves
Size X-Small
For Use With (Application) General Assembly
Glove Cuff Style Knit
Series Y9265
Yarn Gauge 13
Show More Show Less

WARNING: Cancer – www.P65Warnings.ca.gov

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit