Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ FlexTech™ Latex Coated Polyester/Cotton String Knit Gloves

Catalog No. 19148707 Shop All Wells Lamont Industry Group Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Catalog No. 19-148-707

Supplier: Wells Lamont™ Y9240LVP

This item is not returnable. View return policy
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Gloves feature a string knit cotton polyester shell, with a latex coating on the palm for enhanced grip and abrasion resistance.

  • Gray polyester/cotton string-knit shell with blue latex palm coating
  • Latex coating enhances grip capabilities, as well as abrasion/puncture resistance
  • Gray color conceals dirt
  • Flexible and comfortable
  • Packed in individual polybags

Specifications

Coated Area Palm
Construction String Knit
Coating Material Latex
Color Blue, Gray
Exterior Finish Partially Textured
Hand Specific
Material Polycotton
Size Large
For Use With (Application) Canning, Recycling, and General Manufacturing
Glove Cuff Style Knit
Series Y9240-VP
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.