Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Highly Reusable Full-Finger Nylon Glove Liners

Catalog No. 19048646C Shop All Wells Lamont Industry Group Products
Click to view available options
Size:
Small
Medium
Large
X-Large

Highly Reusable Full-Finger Nylon Glove Liners are constructed of 100% nylon and can withstand up to 30 washes without shrinkage.

  • 100% continuous nylon filament construction is thin and comfortable
  • Full finger design offers protection for those with severe glove sensitivities
  • Launderable up to 30 times without any signs of shrinkage
  • Color-coded hems for easy size identification
  • Made in the USA
  • Category II for particulates

Specifications

Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Nylon
Product Type Nylon Glove Liner
Size Large
Finger Style Full
For Use With (Application) Critical Environments, Cleanrooms, and Electronics Assembly
Glove Cuff Style Knit
Hem Color Blue
Series M555
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.