Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ GuardTec3™ Cut-Resistant Gloves with Latex Palm
Shop All Wells Lamont Industry Group Products
Click to view available options
Glove Cuff Color:
Black
Blue
Brown
Green
Red
Size:
Small
Medium
Large
X-Large
2X-Large
For Use With (Application):
Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove
Description
- Features highly cut-resistant fiber for protection against lacerations
- Foam-latex palm coating improves grip
- Knit wrist construction prevents dirt and debris from entering the glove
- Color: Gray
Specifications
Specifications
Color | Gray |
Cut Resistance Rating | ANSI A3 |
Material | Latex, Natural Rubber, Foam |
Product Type | Cut-resistant Gloves |
Size | Small |
For Use With (Application) | Ergonomic benefit of easy grip and prevents dirt and debris from entering the glove |
Glove Cuff Color | Red |
Glove Cuff Style | Knit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction