Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Full-Finger Medical Nylon Glove Liners - Extended Cuff
Shop All Wells Lamont Industry Group Products
Click to view available options
Size:
Small
Medium
Large
Description
- 100% continuous nylon filament creates an ideal barrier between hands and disposable gloves
- Reduces allergies and adverse reactions that can be caused by prolonged exposure to latex
- Packaged in a secure box so that liners remain clean
- Fully launderable up to 20 times without shrinking
- Full finger, extended cuff design
- Non-sterile
- Made in the USA
Specifications
Specifications
Applications | Healthcare |
Coating Material | Uncoated |
Color | White |
Disposable | Reusable |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Nylon |
Product Type | Nylon Glove Liner |
Size | Medium |
Sterility | Non-sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction