Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Scepter™ Antimicrobial Cut-Resistant Medical Glove Liners

Catalog No. 19044178 Shop All Wells Lamont Industry Group Products
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Scepter™ Antimicrobial Cut-Resistant Medical Glove Liners are constructed of stainless steel and polyester to provide comfort, as well as ANSI Cut Level A4 protection. Antimicrobial fibers are latex-free and help to prevent bacterial growth.

  • Stainless steel/polyester fiber blend provides ANSI Cut Level A4 protection
  • Antimicrobial fibers eliminate bacterial growth
  • Moderate tactile sensitivity
  • Color-coded hems for easy size identification
  • Latex-free
  • Made in the USA

Specifications

Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A4
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Stainless Steel, Polyester
Product Type Medical Glove Liner
Size Medium
Sterility Non-sterile
Finger Style Full
For Use With (Application) Medical, Cleanroom, and Semi-Conductor Industries
Glove Cuff Style Knit
Hem Color Orange
Inspected for Medical Use Yes
Packaging Type Individual Polybags
Series M121
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.