Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Aramid Fiber Sleeves

Catalog No. 19013439
Click to view available options
Length (English):
14 in.
16 in.
18 in.
Length (Metric):
35.5 cm
40.6 cm
45.7 cm
Style:
Sleeve with Thumb Hole
Standard Sleeve
Construction:
2-ply
Aramid Fiber with one track
This item is not returnable. View return policy
This item is not returnable. View return policy

These 2 ply Aramid Fiber Sleeves are an ideal solution in providing arm protection and comfort. ANSI A3 rating provides great cut protection, and the material made from para-aramid yarns adds safety with flame-resistant properties. Offer excellent protection against cutting and burn hazards.

  • 2-ply knit sleeve
  • Flame resistant
  • Available with or without a thumb hole
  • ANSI Level 2 Heat Rating
  • ANSI Level 4 Flame Rating
  • ANSI Cut Level A3
  • Made in the USA

Specifications

Product Type Knit Sleeve
Construction 2-ply
Material Aramid Fiber
Color Yellow
Style Standard Sleeve
Disposable Reusable
Cleaning Procedure Remove all large particles. Rinse with cold water then machine wash in warm water with mild detergent or soap for 10 to 15 minutes. Water temperature should not exceed 160°F (71°C). Rinse in cold water. Tumble dry for 10 to 15 minutes, not exceeding 150°F (67°C). Do not use bleach or dry cleaning solution.
Length (English) 16 in.
Length (Metric) 40.6 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.