Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Wells Lamont™ Scepter™ Sterile Antimicrobial M321 Cut-Resistant Gloves
Click to view available options
Size:
Small
Medium
Large
X-Large
Description
- Stainless steel and polyester construction
- Launderable 15 times without shrinking
- Moderate tactile sensitivity
- Antimicrobial
- Sterile and ambidextrous
- Applications: Medical, organ tissue processing and pharmaceutical
- ANSI Cut Level 4
- Made in the USA
Specifications
Specifications
Certifications/Compliance | ASTM F1790-97, ASTM F2992-15 |
Certifications/Compliance (Europe) | Category III |
Coating Material | Uncoated |
Color | White |
Cut Resistance Rating | ANSI A4 |
Hand | Ambidextrous |
Material | Stainless Steel Fiber, Polyester |
Product Type | Cut-resistant Gloves |
Size | Large |
Sterility | Sterile |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction