Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Polypro Coveralls

Catalog No. 19120697E Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Cuff Style:
Elastic
Open
Ankle Style:
Elastic
Open
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Cuff Style Ankle Style
19-120-697E 2X-Large Elastic Elastic
19-142-610F 3X-Large Open Open
19-142-610D X-Large Open Open
19-120-697G 4X-Large Elastic Elastic
19-120-697C Large Elastic Elastic
19-120-697D X-Large Elastic Elastic
19-120-697B Medium Elastic Elastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-120-697E Supplier West Chester Supplier No. 3502/XXL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Offers lightweight protection against dry particulates, dirt, and debris.

  • Available in two styles: open wrists and ankles or elastic wrists and ankles to keep dirt and debris out.
  • Cost-effective standard-weight spunbond polypropylene coverall.
  • Full length zipper front for easy donning.
  • High level of comfort and breathability.

Specifications

Product Type General Purpose Coveralls
Size 2X-Large
Gender Unisex
Material Polypropylene
Color White
Disposable Single-use
Waist Style Loose
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Hood Type Unhooded
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Dust
Latex Free Yes
Model 3502
Size (English) Inseam 30.8 in.
Size (Metric) Inseam 78.2 cm
For Use With (Application) Manufacturing, Pharmaceuticals, Cleaning, Food Processing
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.