Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Polypropylene Disposable Beard Covers

Catalog No. 19121070 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Packaging Quantity:
100/Cs.
1000/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging Quantity
19-121-070 100/Cs.
19-068-442 1000/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-121-070 Supplier West Chester Supplier No. UBC500
This item is not returnable. View return policy

Polypropylene disposable beard cover provides protection against non-hazardous dry particulates.

  • One size fits most
  • Elastic band for flexible fit
  • Latex free
  • White
  • 1000 count
  • Made from component materials that comply with Federal Regulations for Food Contact: 21CFR 170-199

Specifications

Type General Purpose Beard Cover
Size One Size
Material Spunbond Polypropylene
Color White
Disposable Single-use
Latex Free Yes
For Use With (Application) Food Industry, Light Maintenance
Packaging Quantity 100/Cs.
Length (English) 18 in.
Length (Metric) 45.72 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.