Learn More
Description
- Resistant to a wide variety of hazardous chemicals
- Protect against blood born and viral pathogens
- Manufactured with serged seams
- ANSI sized, allowing for freedom of movement
- Low lint
- Full length zipper
Compliance
NFPA 99; ASTM 1670 and 1671
Specifications
Specifications
| Size | Medium |
| Material | Polyethylene, Polypropylene |
| Color | White |
| Waist Style | Elastic |
| Closure Type | Zipper |
| Seam Type | Serged |
| Certifications/Compliance | NFPA 99, U.S. ASTM 1670-1671 |
| Description | Full length zipper front, elastic waistband |
| For Use With (Application) | Painting, asbestos abatement, remediation and Oil handling |
| Length (English) | 36.6 in. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.