Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Posi-Wear™ UB Coveralls

Catalog No. 191304567E Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Length (English):
36.6 in.
38.2 in.
40.2 in.
41.3 in.
43.3 in.
Length (English) Sleeve:
32.7 in.
33.5 in.
35 in.
35.8 in.
37 in.
Width (English) Chest:
24.4 in.
26 in.
27.6 in.
29 in.
31.5 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Length (English) Length (English) Sleeve Width (English) Chest
19-130-4567E 2X-Large 41.3 in. 35.8 in. 29 in.
19-130-4567C Large 38.2 in. 33.5 in. 26 in.
19-130-4567D X-Large 40.2 in. 35 in. 27.6 in.
19-130-4567B Medium 36.6 in. 32.7 in. 24.4 in.
19-130-4567F 3X-Large 43.3 in. 37 in. 31.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-130-4567E Supplier West Chester Supplier No. 3700/XXL
This item is not returnable. View return policy

Provide a high level of protection against chemicals and pathogens

  • Resistant to a wide variety of hazardous chemicals
  • Protect against blood born and viral pathogens
  • Manufactured with serged seams
  • ANSI sized, allowing for freedom of movement
  • Low lint
  • Full length zipper

Compliance

NFPA 99; ASTM 1670 and 1671

Specifications

Size 2X-Large
Material Polyethylene, Polypropylene
Color White
Waist Style Elastic
Closure Type Zipper
Seam Type Serged
Certifications/Compliance NFPA 99, U.S. ASTM 1670-1671
Description Full length zipper front, elastic waistband
For Use With (Application) Painting, asbestos abatement, remediation and Oil handling
Length (English) 41.3 in.
Length (English) Sleeve 35.8 in.
Length (Metric) 104.9 cm
Length (Metric) Sleeve 90.93 cm
Width (English) Chest 29 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.