Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Premium Grain Pigskin Leather Drivers Gloves with Thinsulate™ Lining

Catalog No. 19067581A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-067-581A Small
19-067-581B Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-067-581A Supplier West Chester™ Supplier No. 9940KT/S
This item is not returnable. View return policy

Specifications

Color Yellow
Material Grain Pigskin Leather
Product Type Driver Gloves
Size Small
For Use With (Application) Agriculture, Construction, Foundry, Masonry, and Transportation
Glove Cuff Style Open
Liner Material 3M™ Thinsulate™
Length (English) 9 in.
Length (Metric) 22.8 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.