Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Regular Grade Top Grain Pigskin Leather Drivers Gloves

Catalog No. 19087882A Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-087-882A Small
19-087-882B Medium
19-087-882C Large
19-087-882D X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-087-882A Supplier West Chester™ Supplier No. 994/S
Add to Cart
Add to Cart
This item is not returnable. View return policy

Top grain pigskin leather provides breathability due its porous texture, becomes softer with use, and withstands moisture without stiffening.

  • Top grain leather is smooth, provides durability and dexterity
  • Elastic closure sewn inside glove for safety and comfort
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Straight thumb increases comfort in closed-fisted work
  • Color coded hems for easy size identification
  • Ideal for construction, machine operation, utility work, and many other general applications

Specifications

Color Natural
Grade Regular
Hand Specific
Material Top Grain Pigskin Leather
Product Type Driver Gloves
Size Small
Style Gunn
For Use With (Application) Construction, Machine Operation, Utility Work, and General Applications
Glove Cuff Style Open
Hem Color Red
Series 994
Thumb Style Straight
Length (English) 9 in.
Length (Metric) 22.8 cm
Weight 27.1 lb. (Case)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.