Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester™ Standard Split Cowhide Leather Palm Gloves with Rubberized Cuff

Catalog No. 19821546 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Gender:
Men's
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gender
19-821-546 Men's
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-821-546 Supplier West Chester™ Supplier No. 500L

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Gloves feature a rubberized safety cuff, cotton hem, and wing thumb

  • 2.5 in. rubberized safety cuff
  • Standard split cowhide
  • Wing thumb

Specifications

Cuff Material Rubberized Fabric
Color Gray
Grade Standard
Hand Specific
Material Shoulder Split Cowhide Leather
Product Type Leather Palm Gloves
Size Large
Gender Men's
Glove Cuff Color Green with Pink Stripes
Glove Cuff Style Safety
Hem Color Red
Thumb Style Wing
Length (English) 9.8 in.
Length (Metric) 24.8 cm
Weight 41.1 lb. (Case)
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.