Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Chester Spunbond Polypropylene Lab Coat

Catalog No. 190881684 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Large
X-Large
3X-Large
Thickness:
Standard weight
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Thickness
19-088-1684 Large -
19-088-1685 X-Large Standard weight
19-088-1689 3X-Large -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-088-1684 Supplier West Chester Supplier No. 3511/L
Add to Cart
Add to Cart
This item is not returnable. View return policy

Standard weight lab coats offer breathability and durability. Choose open wrists or elastic wrists style.

  • Lightweight protection against dry particulates
  • Made with durable polypropylene material
  • Snap front closure for secure fit and allows easy donning
  • Collared neck for better air circulation
  • Choose open or elastic wrist style

Specifications

Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Open
Disposable Single-use
Sterility Non-sterile
Resistance Dust
Style Full-length
No. of Pockets None
Collar Type Traditional
Latex Free Yes
Model 3511
Seam Type Serged
For Use With (Application) Manufacturing, Pharmaceuticals, Cleaning, Laboratories
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.